siRNA Details
| Gene Symbol: Hand1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021592.2 | NP_067603.1 | 1 | 707 | ||
| GenBank mRNA | BC097324.1 | - | 1 | 707 | 7460879 | |
| Y08140.1 | - | 1 | 506 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hand1| Gene Symbol: Hand1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021592.2 | NP_067603.1 | 1 | 707 | ||
| GenBank mRNA | BC097324.1 | - | 1 | 707 | 7460879 | |
| Y08140.1 | - | 1 | 506 | |||