siRNA Details
| Gene Symbol: Mcpt8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021598.2 | NP_067609.1 | 523 | |||
| GenBank mRNA | FQ229669.1 | - | 542 | |||
| U67911.1 | - | 524 | ||||
| Gene Symbol: LOC691603 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003751469.3 | XP_003751517.1 | 4 | 584 | ||

Mcpt8
Genomic Map