siRNA Details
| Gene Symbol: Cml1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021668.3 | NP_067700.2 | 2 | 793 | ||
| XM_006236809.2 | XP_006236871.1 | 694 | ||||
| GenBank mRNA | AF185569.1 | - | 735 | |||
| BC078836.1 | - | 774 | 7106831 | |||
| Gene Symbol: LOC103690120 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006236788.2 | XP_006236850.1 | 1 | 694 | ||
| XM_008763110.1 | XP_008761332.1 | 2 | 774 | |||

Cml1
Genomic Map