siRNA Details
| Gene Symbol: F13a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021698.2 | NP_067730.2 | 4 | 482 | ||
| GenBank mRNA | Y12502.1 | - | 4 | 449 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

F13a1| Gene Symbol: F13a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021698.2 | NP_067730.2 | 4 | 482 | ||
| GenBank mRNA | Y12502.1 | - | 4 | 449 | ||