siRNA Details
| Gene Symbol: Mapk12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021746.1 | NP_068514.1 | 9 | 896 | ||
| XM_008765781.1 | XP_008764003.1 | 9 | 625 | |||
| GenBank mRNA | BC081950.1 | - | 9 | 875 | ||
| BC127549.1 | - | 2 | 53 | |||
| X96488.1 | - | 9 | 896 | |||

Mapk12
Genomic Map