siRNA Details
| Gene Symbol: Plrg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021757.2 | NP_068525.1 | 9, 10 | 938 | ||
| GenBank mRNA | AF128241.1 | - | 8, 9 | 845 | ||
| BC087742.1 | - | 9, 10 | 938 | 7314208 | ||
| FQ215141.1 | - | 9, 10 | 959 | |||

Plrg1
Genomic Map