siRNA Details
| Gene Symbol: Lypd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021759.1 | NP_068527.1 | 2 | 211 | ||
| GenBank mRNA | AJ001043.1 | - | 2 | 211 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lypd3| Gene Symbol: Lypd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021759.1 | NP_068527.1 | 2 | 211 | ||
| GenBank mRNA | AJ001043.1 | - | 2 | 211 | ||