siRNA Details
| Gene Symbol: Gabra6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021841.1 | NP_068613.1 | 2 | 81 | ||
| XM_006246128.2 | XP_006246190.1 | 2 | 260 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gabra6| Gene Symbol: Gabra6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021841.1 | NP_068613.1 | 2 | 81 | ||
| XM_006246128.2 | XP_006246190.1 | 2 | 260 | |||