siRNA Details
| Gene Symbol: Kcnt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021853.1 | NP_068625.1 | 1, 2 | 163 | ||
| XM_006233697.2 | XP_006233759.1 | 1, 2 | 471 | |||
| XM_006233698.2 | XP_006233760.1 | 1, 2 | 471 | |||
| XM_006233699.2 | XP_006233761.1 | 1, 2 | 471 | |||
| XM_006233701.2 | XP_006233763.1 | 1, 2 | 471 | |||
| XM_006233703.2 | XP_006233765.1 | 1, 2 | 471 | |||
| XM_006233706.2 | XP_006233768.1 | 1, 2 | 471 | |||
| GenBank mRNA | AF089730.1 | - | 1, 2 | 163 | ||

Kcnt1
Genomic Map