siRNA Details
| Gene Symbol: Tsc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021854.1 | NP_068626.1 | 1, 2 | 98 | ||
| XM_006233843.2 | XP_006233905.1 | 3, 4 | 552 | |||
| XM_006233844.2 | XP_006233906.1 | 3, 4 | 523 | |||
| XM_006233845.2 | XP_006233907.1 | 3, 4 | 374 | |||
| XM_006233846.2 | XP_006233908.1 | 3, 4 | 2639 | |||
| XM_006233847.2 | XP_006233909.1 | 3, 4 | 860 | |||
| XM_006233849.2 | XP_006233911.1 | 3, 4 | 552 | |||
| GenBank mRNA | AB011821.1 | - | 1, 2 | 98 | ||

Tsc1
Genomic Map