siRNA Details
| Gene Symbol: Rhox5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022175.1 | NP_071511.1 | 2 | 116 | ||
| XM_006257472.2 | XP_006257534.1 | 3 | 565 | |||
| XM_006257473.2 | XP_006257535.1 | 3 | 181 | |||
| XM_006257475.1 | XP_006257537.1 | 4 | 309 | |||
| XM_008773400.1 | XP_008771622.1 | 4 | 460 | |||
| GenBank mRNA | DQ058653.1 | - | 2 | 116 | ||
| FQ228987.1 | - | 3 | 199 | |||
| S82627.1 | - | 2 | 101 | |||

Rhox5
Genomic Map