siRNA Details
| Gene Symbol: Myo5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022178.1 | NP_071514.1 | 135 | |||
| GenBank mRNA | AB035736.1 | - | 135 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Myo5a| Gene Symbol: Myo5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022178.1 | NP_071514.1 | 135 | |||
| GenBank mRNA | AB035736.1 | - | 135 | |||