siRNA Details
| Gene Symbol: Hnf4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022180.2 | NP_071516.2 | 1 | 244 | ||
| XM_006235515.2 | XP_006235577.1 | 1 | 239 | |||
| XM_006235516.2 | XP_006235578.1 | 1 | 239 | |||
| GenBank mRNA | CK483260.1 | - | 1 | 73 | ||
| D10554.1 | - | 1 | 120 | |||
| EF193392.1 | - | 1 | 46 | |||
| FM042560.1 | - | 1 | 249 | |||
| X57133.1 | - | 1 | 135 | |||

Hnf4a
Genomic Map