siRNA Details
| Gene Symbol: Amph | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022217.1 | NP_071553.1 | 2 | 98 | ||
| GenBank mRNA | Y13381.1 | - | 2 | 98 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Amph| Gene Symbol: Amph | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022217.1 | NP_071553.1 | 2 | 98 | ||
| GenBank mRNA | Y13381.1 | - | 2 | 98 | ||