siRNA Details
| Gene Symbol: Tsn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021762.1 | NP_068530.1 | 2, 3 | 270 | ||
| XM_008769496.1 | XP_008767718.1 | 2, 3 | 289 | |||
| GenBank mRNA | AF262356.1 | - | 2, 3 | 270 | ||
| AY325258.1 | - | 345 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tsn| Gene Symbol: Tsn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021762.1 | NP_068530.1 | 2, 3 | 270 | ||
| XM_008769496.1 | XP_008767718.1 | 2, 3 | 289 | |||
| GenBank mRNA | AF262356.1 | - | 2, 3 | 270 | ||
| AY325258.1 | - | 345 | ||||