siRNA Details
| Gene Symbol: Trpc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021771.2 | NP_068539.2 | 2 | 828 | ||
| XM_006232282.2 | XP_006232344.2 | 2 | 993 | |||
| XM_008760941.1 | XP_008759163.1 | 2 | 993 | |||
| GenBank mRNA | JN160741.1 | - | 2 | 589 | ||

Trpc3
Genomic Map