siRNA Details
| Gene Symbol: Jun | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021835.3 | NP_068607.1 | 1 | 1787 | ||
| GenBank mRNA | BC078738.1 | - | 1 | 1024 | 7124370 | |
| X17163.1 | - | 1 | 1281 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Jun| Gene Symbol: Jun | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021835.3 | NP_068607.1 | 1 | 1787 | ||
| GenBank mRNA | BC078738.1 | - | 1 | 1024 | 7124370 | |
| X17163.1 | - | 1 | 1281 | |||