siRNA Details
| Gene Symbol: Junb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021836.2 | NP_068608.2 | 1 | 1166 | ||
| GenBank mRNA | BC061862.1 | - | 1 | 1166 | 5597977 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Junb| Gene Symbol: Junb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021836.2 | NP_068608.2 | 1 | 1166 | ||
| GenBank mRNA | BC061862.1 | - | 1 | 1166 | 5597977 | |