siRNA Details
| Gene Symbol: Crx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021855.1 | NP_068627.1 | 2, 3 | 260 | ||
| XM_006228354.1 | XP_006228416.1 | 3, 4 | 444 | |||
| GenBank mRNA | AB021129.1 | - | 2, 3 | 260 | ||
| AF135838.1 | - | 2, 3 | 260 | |||

Crx
Genomic Map