siRNA Details
| Gene Symbol: Pik3r2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022185.2 | NP_071521.2 | 8, 9 | 1440 | ||
| XM_008771096.1 | XP_008769318.1 | 7, 8 | 2600 | |||
| GenBank mRNA | BC089805.1 | - | 8, 9 | 1440 | 7381613 | |
| D64046.1 | - | 7, 8 | 1000 | |||

Pik3r2
Genomic Map