siRNA Details
| Gene Symbol: Ppp2r2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022209.3 | NP_071545.2 | 4 | 598 | ||
| GenBank mRNA | AY251277.1 | - | 4 | 550 | ||
| BC078834.1 | - | 7 | 807 | 7114294 | ||
| D14421.1 | - | 4 | 373 | |||
| D38260.1 | - | 4 | 393 | |||
| FQ212581.1 | - | 2 | 118 | |||

Ppp2r2b
Genomic Map