siRNA Details
| Gene Symbol: Max | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022210.1 | NP_071546.1 | 5 | 320 | ||
| XM_006240248.2 | XP_006240310.1 | 4 | 519 | |||
| XM_008764846.1 | XP_008763068.1 | 5 | 1309 | |||
| GenBank mRNA | BC099778.1 | - | 6 | 567 | 7933028 | |
| D14447.1 | - | 5 | 320 | |||
| D14448.1 | - | 4 | 293 | |||

Max
Genomic Map