siRNA Details
| Gene Symbol: Pgrmc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021766.1 | NP_068534.1 | 2 | 513 | ||
| GenBank mRNA | AF163321.1 | - | 2 | 476 | ||
| AJ005837.1 | - | 2 | 490 | |||
| BC062073.1 | - | 2 | 479 | 5599449 | ||
| U63315.1 | - | 2 | 513 | |||

Pgrmc1
Genomic Map