siRNA Details
| Gene Symbol: Hspa2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021863.3 | NP_068635.1 | 1 | 1828 | ||
| XM_006240247.2 | XP_006240309.1 | 2 | 1805 | |||
| GenBank mRNA | BC081803.1 | - | 2 | 1799 | 7112083 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hspa2| Gene Symbol: Hspa2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021863.3 | NP_068635.1 | 1 | 1828 | ||
| XM_006240247.2 | XP_006240309.1 | 2 | 1805 | |||
| GenBank mRNA | BC081803.1 | - | 2 | 1799 | 7112083 | |