siRNA Details
Gene Symbol: Rhox5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022175.1 | NP_071511.1 | 3, 4 | 493 | ||
GenBank mRNA | DQ058653.1 | - | 3, 4 | 493 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Rhox5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022175.1 | NP_071511.1 | 3, 4 | 493 | ||
GenBank mRNA | DQ058653.1 | - | 3, 4 | 493 |