siRNA Details
| Gene Symbol: Clcn4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022198.2 | NP_071534.2 | 11 | 1894 | ||
| XM_006256828.2 | XP_006256890.1 | 11 | 2197 | |||
| GenBank mRNA | BC092209.1 | - | 11 | 1894 | 7315063 | |
| FQ210965.1 | - | 11 | 1933 | |||
| Z36944.1 | - | 9 | 1602 | |||

Clcn4
Genomic Map