siRNA Details
| Gene Symbol: Unc5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022207.1 | NP_071543.1 | 12 | 1794 | ||
| XM_006256423.2 | XP_006256485.1 | 11 | 2157 | |||
| GenBank mRNA | U87306.1 | - | 12 | 1794 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Unc5b| Gene Symbol: Unc5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022207.1 | NP_071543.1 | 12 | 1794 | ||
| XM_006256423.2 | XP_006256485.1 | 11 | 2157 | |||
| GenBank mRNA | U87306.1 | - | 12 | 1794 | ||