siRNA Details
| Gene Symbol: Nmu | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022239.2 | NP_071575.1 | 3, 4 | 345 | ||
| XM_006250872.2 | XP_006250934.1 | 3, 4 | 1165 | |||
| GenBank mRNA | AJ510132.1 | - | 2, 3 | 104 | ||
| M94555.1 | - | 3, 4 | 345 | |||

Nmu
Genomic Map