siRNA Details
Gene Symbol: Dusp12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022248.1 | NP_071584.1 | 3 | 581 | ||
GenBank mRNA | AF217233.1 | - | 3 | 581 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Dusp12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022248.1 | NP_071584.1 | 3 | 581 | ||
GenBank mRNA | AF217233.1 | - | 3 | 581 |