siRNA Details
| Gene Symbol: Cd244 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022259.1 | NP_071595.1 | 2 | 409 | ||
| XM_006250259.2 | XP_006250321.1 | 2 | 1788 | |||
| GenBank mRNA | AF156988.1 | - | 2 | 469 | ||
| AF156989.1 | - | 2 | 409 | |||
| AF209406.1 | - | 2 | 406 | |||

Cd244
Genomic Map