siRNA Details
| Gene Symbol: Bspry | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022261.2 | NP_071597.2 | 3 | 556 | ||
| GenBank mRNA | AF245225.1 | - | 1 | 83 | ||
| BC062051.1 | - | 3 | 504 | |||
| BC095901.1 | - | 3 | 207 | |||
| BC111380.1 | - | 2 | 110 | |||
| BC166587.1 | - | 3 | 556 | 7937509 | ||
| DN935023.1 | - | 3 | 586 | |||

Bspry
Genomic Map