siRNA Details
| Gene Symbol: Kit | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022264.1 | NP_071600.1 | 2 | 273 | ||
| XM_006250909.2 | XP_006250971.1 | 2 | 307 | |||
| GenBank mRNA | AF228307.1 | - | 2 | 250 | ||
| AF228308.1 | - | 2 | 247 | |||
| AF228309.1 | - | 2 | 250 | |||
| AF228310.1 | - | 2 | 250 | |||
| AF228311.1 | - | 2 | 250 | |||
| AF296694.1 | - | 2 | 257 | |||
| D12524.1 | - | 2 | 273 | |||
| EU247827.1 | - | 2 | 257 | |||
| EU247828.1 | - | 2 | 257 | |||

Kit
Genomic Map