siRNA Details
| Gene Symbol: Ctgf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022266.2 | NP_071602.1 | 4, 5 | 967 | ||
| GenBank mRNA | AB023068.1 | - | 4, 5 | 955 | ||
| AF079531.1 | - | 1, 2 | 101 | |||
| AF120275.1 | - | 4, 5 | 967 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ctgf| Gene Symbol: Ctgf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022266.2 | NP_071602.1 | 4, 5 | 967 | ||
| GenBank mRNA | AB023068.1 | - | 4, 5 | 955 | ||
| AF079531.1 | - | 1, 2 | 101 | |||
| AF120275.1 | - | 4, 5 | 967 | |||