siRNA Details
| Gene Symbol: Vps33b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022286.2 | NP_071622.1 | 2 | 430 | ||
| GenBank mRNA | BC081707.1 | - | 2 | 430 | 7128590 | |
| U35245.1 | - | 2 | 145 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vps33b| Gene Symbol: Vps33b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022286.2 | NP_071622.1 | 2 | 430 | ||
| GenBank mRNA | BC081707.1 | - | 2 | 430 | 7128590 | |
| U35245.1 | - | 2 | 145 | |||