siRNA Details
| Gene Symbol: A4galt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022240.2 | NP_071576.1 | 1 | 287 | ||
| XM_006242115.2 | XP_006242177.1 | 3 | 672 | |||
| XM_006242117.2 | XP_006242179.1 | 3 | 548 | |||
| XM_006242118.2 | XP_006242180.1 | 2 | 772 | |||
| XM_006242120.1 | XP_006242182.1 | 3 | 435 | |||
| GenBank mRNA | AF248544.1 | - | 1 | 301 | ||
| BC097323.1 | - | 1 | 287 | 7459302 | ||

A4galt
Genomic Map