siRNA Details
| Gene Symbol: Masp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022257.1 | NP_071593.1 | 2, 3 | 357 | ||
| XM_006248526.2 | XP_006248588.1 | 2, 3 | 601 | |||
| XM_008768804.1 | XP_008767026.1 | 2, 3 | 595 | |||
| GenBank mRNA | AF004661.1 | - | 1, 2 | 227 | ||
| AJ277423.1 | - | 1, 2 | 253 | |||
| AJ457084.1 | - | 2, 3 | 357 | |||
| AJ487622.1 | - | 1, 2 | 153 | |||
| BC085685.1 | - | 2, 3 | 450 | 7191018 | ||

Masp1
Genomic Map