siRNA Details
| Gene Symbol: Ctgf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022266.2 | NP_071602.1 | 4, 5 | 982 | ||
| GenBank mRNA | AB023068.1 | - | 4, 5 | 970 | ||
| AF079531.1 | - | 1, 2 | 116 | |||
| AF120275.1 | - | 4, 5 | 982 | |||
| AY596447.1 | - | 1 | 1543 | |||
| BC072503.1 | - | 4 | 1351 | 7098774 | ||

Ctgf
Genomic Map