siRNA Details
| Gene Symbol: Fbxl20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022272.1 | NP_071608.1 | 5 | 279 | ||
| XM_006247421.2 | XP_006247483.1 | 5 | 581 | |||
| XM_008768110.1 | XP_008766332.1 | 5 | 583 | |||
| XM_008768111.1 | XP_008766333.1 | 5 | 618 | |||
| GenBank mRNA | AF182443.1 | - | 5 | 279 | ||

Fbxl20
Genomic Map