siRNA Details
Gene Symbol: FOXE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004473.3 | NP_004464.2 | 1 | 2640 | ||
GenBank mRNA | U89995.1 | - | 1 | 2677 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: FOXE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004473.3 | NP_004464.2 | 1 | 2640 | ||
GenBank mRNA | U89995.1 | - | 1 | 2677 |