siRNA Details
| Gene Symbol: Acvrl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022441.2 | NP_071886.1 | 9 | 1460 | ||
| XM_006242303.2 | XP_006242365.1 | 10 | 1861 | |||
| XM_006242304.2 | XP_006242366.1 | 10 | 2465 | |||
| XM_006242305.2 | XP_006242367.1 | 9 | 2480 | |||
| XM_006242307.2 | XP_006242369.1 | 9 | 2350 | |||
| XM_008765673.1 | XP_008763895.1 | 10 | 2524 | |||
| GenBank mRNA | BC083173.1 | - | 9 | 1460 | 7098096 | |
| FQ223580.1 | - | 9 | 1502 | |||
| L36088.1 | - | 10 | 1875 | |||

Acvrl1
Genomic Map