siRNA Details
| Gene Symbol: Aldh9a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022273.2 | NP_071609.2 | 6 | 817 | ||
| GenBank mRNA | AF170918.1 | - | 6 | 804 | ||
| BC074019.1 | - | 6 | 817 | 7104823 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Aldh9a1| Gene Symbol: Aldh9a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022273.2 | NP_071609.2 | 6 | 817 | ||
| GenBank mRNA | AF170918.1 | - | 6 | 804 | ||
| BC074019.1 | - | 6 | 817 | 7104823 | ||