siRNA Details
| Gene Symbol: Mre11a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022279.1 | NP_071615.1 | 5 | 500 | ||
| XM_006242532.2 | XP_006242594.1 | 5 | 595 | |||
| XM_006242533.1 | XP_006242595.1 | 5 | 589 | |||
| XM_006242534.2 | XP_006242596.1 | 6 | 671 | |||
| GenBank mRNA | AF218574.1 | - | 5 | 500 | ||

Mre11a
Genomic Map