siRNA Details
| Gene Symbol: Lrat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022280.2 | NP_071616.1 | 2 | 906 | ||
| GenBank mRNA | AF255060.2 | - | 2 | 906 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lrat| Gene Symbol: Lrat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022280.2 | NP_071616.1 | 2 | 906 | ||
| GenBank mRNA | AF255060.2 | - | 2 | 906 | ||