siRNA Details
| Gene Symbol: Kcnk13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022293.1 | NP_071629.1 | 2 | 1069 | ||
| XM_006240447.2 | XP_006240509.1 | 3 | 3867 | |||
| GenBank mRNA | AF287301.1 | - | 2 | 1069 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kcnk13| Gene Symbol: Kcnk13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022293.1 | NP_071629.1 | 2 | 1069 | ||
| XM_006240447.2 | XP_006240509.1 | 3 | 3867 | |||
| GenBank mRNA | AF287301.1 | - | 2 | 1069 | ||