siRNA Details
| Gene Symbol: Prkcz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022507.1 | NP_071952.1 | 6, 7 | 700 | ||
| XM_006239535.2 | XP_006239597.1 | 3, 4 | 3195 | |||
| XM_006239536.2 | XP_006239598.1 | 3, 4 | 2401 | |||
| XM_008764337.1 | XP_008762559.1 | 6, 7 | 601 | |||
| GenBank mRNA | J04532.1 | - | 6, 7 | 700 | ||
| M18332.1 | - | 3, 4 | 253 | |||
| U27191.1 | - | 3, 4 | 465 | |||

Prkcz
Genomic Map