siRNA Details
Gene Symbol: FOXK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004514.3 | NP_004505.2 | 3 | 914 | ||
GenBank mRNA | U58196.1 | - | 3 | 925 | ||
U58197.1 | - | 3 | 925 | |||
U58198.1 | - | 3 | 925 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: FOXK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004514.3 | NP_004505.2 | 3 | 914 | ||
GenBank mRNA | U58196.1 | - | 3 | 925 | ||
U58197.1 | - | 3 | 925 | |||
U58198.1 | - | 3 | 925 |