siRNA Details
Gene Symbol: Gpx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022525.3 | NP_071970.2 | 2 | 229 | ||
GenBank mRNA | BC062227.1 | - | 2 | 205 | 6921653 | |
CK480346.1 | - | 2 | 229 | |||
D00680.1 | - | 2 | 294 | |||
FQ221037.1 | - | 2 | 201 | |||
FQ221279.1 | - | 2 | 182 | |||
FQ221454.1 | - | 2 | 201 | |||
FQ228485.1 | - | 2 | 201 | |||
FQ228545.1 | - | 2 | 255 | |||
FQ228679.1 | - | 2 | 201 | |||
FQ228689.1 | - | 2 | 201 | |||
FQ228690.1 | - | 2 | 214 | |||
FQ228694.1 | - | 2 | 181 | |||
FQ228697.1 | - | 2 | 201 | |||
FQ228699.1 | - | 2 | 201 | |||
FQ228705.1 | - | 2 | 199 | |||
FQ228712.1 | - | 2 | 201 | |||
FQ228719.1 | - | 2 | 201 | |||
FQ228737.1 | - | 2 | 201 | |||
FQ228751.1 | - | 2 | 201 | |||
FQ228788.1 | - | 2 | 201 | |||
FQ228802.1 | - | 2 | 201 | |||
FQ228803.1 | - | 2 | 198 | |||
FQ228804.1 | - | 2 | 201 | |||
FQ228807.1 | - | 2 | 202 | |||
FQ228808.1 | - | 2 | 201 | |||
FQ228824.1 | - | 2 | 201 | |||
FQ228834.1 | - | 2 | 203 | |||
FQ228835.1 | - | 2 | 201 | |||
FQ228845.1 | - | 2 | 201 | |||
FQ228866.1 | - | 2 | 201 | |||
FQ228878.1 | - | 2 | 201 | |||
FQ228879.1 | - | 2 | 201 | |||
FQ228886.1 | - | 2 | 201 | |||
FQ228899.1 | - | 2 | 201 | |||
FQ228911.1 | - | 2 | 201 | |||
FQ228914.1 | - | 2 | 201 | |||
FQ228921.1 | - | 2 | 201 | |||
FQ228934.1 | - | 2 | 203 | |||
FQ228945.1 | - | 2 | 201 | |||
FQ228947.1 | - | 2 | 201 | |||
FQ228960.1 | - | 2 | 201 | |||
FQ228965.1 | - | 2 | 201 | |||
FQ228968.1 | - | 2 | 201 | |||
FQ228969.1 | - | 2 | 200 | |||
FQ228970.1 | - | 2 | 201 | |||
FQ228976.1 | - | 2 | 201 | |||
FQ229118.1 | - | 2 | 201 | |||
FQ229125.1 | - | 2 | 201 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
