siRNA Details
| Gene Symbol: Araf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033663.2 | NP_001028835.1 | 3, 4 | 317 | ||
| NM_022532.2 | NP_071977.1 | 3, 4 | 317 | |||
| XM_008773036.1 | XP_008771258.1 | 3, 4 | 1059 | |||
| GenBank mRNA | AB158253.1 | - | 2, 3 | 213 | ||
| BC060568.1 | - | 357 | ||||
| CO572305.1 | - | 3, 4 | 317 | |||
| X06942.1 | - | 2, 3 | 284 | |||

Araf
Genomic Map