siRNA Details
| Gene Symbol: Ppib | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022536.2 | NP_071981.1 | 1 | 120 | ||
| GenBank mRNA | AF071225.1 | - | 1 | 120 | ||
| BC061971.1 | - | 1 | 149 | 5623342 | ||
| FQ211358.1 | - | 1 | 124 | |||
| FQ212031.1 | - | 1 | 149 | |||
| FQ219792.1 | - | 1 | 158 | |||
| FQ219926.1 | - | 1 | 170 | |||
| FQ221020.1 | - | 1 | 173 | |||
| FQ221248.1 | - | 1 | 168 | |||
| FQ222696.1 | - | 1 | 282 | |||
| FQ223162.1 | - | 1 | 167 | |||
| FQ228087.1 | - | 1 | 163 | |||
| FQ229174.1 | - | 1 | 170 | |||
| FQ229214.1 | - | 1 | 164 | |||
| FQ230004.1 | - | 174 | ||||
| FQ231656.1 | - | 1 | 163 | |||
| FQ232077.1 | - | 1 | 159 | |||
| FQ232920.1 | - | 1 | 173 | |||

Ppib
Genomic Map