siRNA Details
| Gene Symbol: Ccdc80 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022543.2 | NP_071988.2 | 2 | 1694 | ||
| GenBank mRNA | AF223677.1 | - | 2 | 1801 | ||
| AY548105.1 | - | 2 | 1694 | |||
| BC070926.1 | - | 1 | 1032 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc80| Gene Symbol: Ccdc80 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022543.2 | NP_071988.2 | 2 | 1694 | ||
| GenBank mRNA | AF223677.1 | - | 2 | 1801 | ||
| AY548105.1 | - | 2 | 1694 | |||
| BC070926.1 | - | 1 | 1032 | |||